Assay Details
Gene Symbol: Bicd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108653.1 | NP_001102123.1 | 2 - 3 | 857 | 134 | |
XM_006237703.2 | XP_006237765.1 | 2 - 3 | 837 | 134 | ||
XM_006237705.2 | XP_006237767.1 | 2 - 3 | 837 | 134 | ||
XM_006237706.2 | XP_006237768.1 | 2 - 3 | 837 | 134 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
viral process
protein localization to organelle
stress granule assembly
positive regulation of receptor-mediated endocytosis
minus-end-directed organelle transport along microtubule
microtubule anchoring at microtubule organizing center
negative regulation of phospholipase C activity
regulation of proteinase activated receptor activity
negative regulation of phospholipase C-activating G protein-coupled receptor signaling pathway
cytoskeletal anchor activity
proteinase activated receptor binding
dynactin binding
dynein complex binding
viral process
protein localization to organelle
stress granule assembly
positive regulation of receptor-mediated endocytosis
minus-end-directed organelle transport along microtubule
microtubule anchoring at microtubule organizing center
negative regulation of phospholipase C activity
regulation of proteinase activated receptor activity
negative regulation of phospholipase C-activating G protein-coupled receptor signaling pathway
cytoskeletal anchor activity
proteinase activated receptor binding
dynactin binding
dynein complex binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P109387 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 808865 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 609763 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 416945 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149340 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380530 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 778847 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 769462 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908891 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 359711 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 45652 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 353558 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698820 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 515174 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 551207 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873258 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 845915 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 722457 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 724417 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640321 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 269561 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27902 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 45011 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 342698 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27208 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 772544 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706686 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 324913 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 384200 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 11676 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 532993 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 932657 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 826015 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687997 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908284 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 287251 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 399429 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 464764 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 197767 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 495634 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 1288 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6846 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613820 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965711 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617076 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797578 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634766 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 943359 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 666317 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 533840 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588590 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 926070 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 929116 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872986 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 905495 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 223355 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147503 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 189484 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 490127 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 570553 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 533851 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535910 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143216 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947072 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907599 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434155 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640165 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 723528 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839681 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 216033 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 430071 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 282490 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144949 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 299751 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419734 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63061 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 480084 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 775767 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 663258 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574937 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 811716 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 476154 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 838183 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 425492 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 547337 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375938 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 119470 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 257699 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 703990 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 104660 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 295646 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 957658 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910052 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 307013 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 824131 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47696 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177548 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 25223 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16922 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26136 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 146925 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 124635 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 111290 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151736 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 103860 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 60088 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 23626 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 80483 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 172446 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 173485 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 56877 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 73345 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 158734 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P109387 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P109387 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P109387 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P128693 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 73814 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 25798 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 4275 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 61694 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 11690 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 42710 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 62205 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 40595 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10859787 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10859786 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10859785 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10859783 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10859790 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10859791 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10859788 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10859782 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10859784 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10859781 | - | - |
Affymetrix Rat Clariom S Assay | PSR0400015044.rn.1 | TC0400002100.rn.1 | RTA TC |
Affymetrix Rat Gene 1.0 ST Array | 10859781 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21658126 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1352661 | - | - |
