Assay Details
Gene Symbol: Amn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008333.2 | NP_001008334.2 | 2 - 3 | 222 | 117 | |
XM_006237700.1 | XP_006237762.1 | 2 - 3 | 708 | 117 | ||
GenBank mRNA | BC086357.1 | - | 2 - 3 | 192 | 117 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC086357.1, XM_006237700.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
