Assay Details
Gene Symbol: Amn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008333.2 | NP_001008334.2 | 3 - 4 | 364 | 145 | |
XM_006237700.1 | XP_006237762.1 | 3 - 4 | 850 | 145 | ||
GenBank mRNA | BC086357.1 | - | 3 - 4 | 334 | 145 | |
BG672304.1 | - | 3 - 4 | 364 | 145 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
