Assay Details
Gene Symbol: Eno2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139325.3 | NP_647541.1 | 8 - 9 | 935 | 65 | |
XM_006237330.2 | XP_006237392.1 | 8 - 9 | 1221 | 65 | ||
GenBank mRNA | AF019973.1 | - | 8 - 9 | 935 | 65 | |
BC060310.1 | - | 8 - 9 | 926 | 65 | ||
M11931.1 | - | 8 - 9 | 935 | 65 |
Gene Symbol: LOC100911625 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750657.3 | XP_003750705.1 | 8 - 9 | 991 | 65 | |
XM_006244636.2 | XP_006244698.1 | 8 - 9 | 1223 | 65 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
