Assay Details
Gene Symbol: Phc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107886.1 | NP_001101356.1 | 10 - 11 | 2420 | 76 | |
XM_006237300.2 | XP_006237362.1 | 11 - 12 | 2445 | 76 | ||
XM_006237302.2 | XP_006237364.1 | 9 - 10 | 2343 | 76 | ||
XM_006237303.2 | XP_006237365.1 | 10 - 11 | 2285 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
