Assay Details
Gene Symbol: Adipor2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037979.1 | NP_001033068.1 | 5 - 6 | 852 | 135 | |
XM_006237183.1 | XP_006237245.1 | 6 - 7 | 1158 | 135 | ||
XM_006237184.2 | XP_006237246.1 | 6 - 7 | 1029 | 135 | ||
XM_006237185.2 | XP_006237247.1 | 6 - 7 | 1044 | 135 | ||
XM_006237186.2 | XP_006237248.1 | 6 - 7 | 1173 | 135 | ||
GenBank mRNA | AY724514.1 | - | 6 - 7 | 1112 | 135 | |
DQ148392.1 | - | 5 - 6 | 852 | 135 | ||
FQ232508.1 | - | 6 - 7 | 1131 | 135 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
