Assay Details
Gene Symbol: Cd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053018.1 | NP_444177.1 | 3 - 4 | 330 | 61 | |
GenBank mRNA | BC161804.1 | - | 3 - 4 | 305 | 61 | |
CF111078.1 | - | 3 - 4 | 339 | 61 | ||
FQ212497.1 | - | 3 - 4 | 354 | 61 | ||
FQ213039.1 | - | 3 - 4 | 379 | 61 | ||
FQ213302.1 | - | 3 - 4 | 348 | 61 | ||
FQ215636.1 | - | 3 - 4 | 342 | 61 | ||
FQ216402.1 | - | 3 - 4 | 379 | 61 | ||
FQ216770.1 | - | 3 - 4 | 366 | 61 | ||
FQ219799.1 | - | 3 - 4 | 353 | 61 | ||
FQ220476.1 | - | 3 - 4 | 354 | 61 | ||
FQ224453.1 | - | 3 - 4 | 354 | 61 | ||
FQ229965.1 | - | 3 - 4 | 401 | 61 | ||
KC162232.1 | - | 269 | 61 | |||
KC162233.1 | - | 2 - 3 | 316 | 61 | ||
X76489.1 | - | 3 - 4 | 269 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
