Assay Details
Gene Symbol: Cd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053018.1 | NP_444177.1 | 4 - 5 | 401 | 129 | |
GenBank mRNA | BC161804.1 | - | 4 - 5 | 376 | 129 | |
CF111078.1 | - | 4 - 5 | 410 | 129 | ||
FQ212497.1 | - | 4 - 5 | 425 | 129 | ||
FQ213039.1 | - | 4 - 5 | 450 | 129 | ||
FQ213302.1 | - | 4 - 5 | 419 | 129 | ||
FQ215636.1 | - | 4 - 5 | 413 | 129 | ||
FQ216402.1 | - | 4 - 5 | 450 | 129 | ||
FQ216770.1 | - | 4 - 5 | 437 | 129 | ||
FQ219799.1 | - | 4 - 5 | 424 | 129 | ||
FQ220476.1 | - | 4 - 5 | 425 | 129 | ||
FQ224453.1 | - | 4 - 5 | 425 | 129 | ||
FQ229965.1 | - | 4 - 5 | 472 | 129 | ||
X76489.1 | - | 4 - 5 | 340 | 129 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
