Assay Details
Gene Symbol: Cd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053018.1 | NP_444177.1 | 6 - 7 | 595 | 126 | |
GenBank mRNA | BC161804.1 | - | 6 - 7 | 570 | 126 | |
BG670145.1 | - | 2 - 3 | 187 | 126 | ||
BQ202980.1 | - | 2 - 3 | 126 | |||
CF111078.1 | - | 6 - 7 | 604 | 126 | ||
FQ212497.1 | - | 6 - 7 | 619 | 126 | ||
FQ213039.1 | - | 6 - 7 | 644 | 126 | ||
FQ213302.1 | - | 6 - 7 | 613 | 126 | ||
FQ215636.1 | - | 6 - 7 | 607 | 126 | ||
FQ216402.1 | - | 6 - 7 | 644 | 126 | ||
FQ229965.1 | - | 6 - 7 | 666 | 126 | ||
FQ230141.1 | - | 6 - 7 | 681 | 126 | ||
X76489.1 | - | 6 - 7 | 534 | 126 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
