Assay Details
Gene Symbol: Tcf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006246362.2 | XP_006246424.1 | 9 - 10 | 1356 | 93 | |
XM_006246363.2 | XP_006246425.1 | 9 - 10 | 1366 | 93 | ||
XM_006246364.2 | XP_006246426.1 | 9 - 10 | 1360 | 93 | ||
XM_006246365.2 | XP_006246427.1 | 8 - 9 | 1263 | 93 | ||
XM_008767741.1 | XP_008765963.1 | 9 - 10 | 3180 | 93 | ||
GenBank mRNA | FQ233211.1 | - | 3 - 4 | 323 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
