Assay Details
Gene Symbol: Dhrs11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014119.2 | NP_001014141.2 | 3 - 4 | 559 | 135 | |
XM_006247045.2 | XP_006247107.1 | 3 - 4 | 680 | 135 | ||
GenBank mRNA | BC088444.1 | - | 3 - 4 | 553 | 135 | |
DY311021.1 | - | 3 - 4 | 559 | 135 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
