Assay Details
Gene Symbol: Dhrs11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014119.2 | NP_001014141.2 | 4 - 5 | 691 | 65 | |
GenBank mRNA | BC088444.1 | - | 4 - 5 | 685 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
