Assay Details
Gene Symbol: Dhrs11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014119.2 | NP_001014141.2 | 6 - 7 | 848 | 72 | |
XM_008768069.1 | XP_008766291.1 | 5 - 6 | 867 | 72 | ||
GenBank mRNA | BC088444.1 | - | 6 - 7 | 842 | 72 | |
FQ233234.1 | - | 1 - 2 | 57 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
