Assay Details
Gene Symbol: Psmd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008281.1 | NP_001008282.1 | 8 - 9 | 1347 | 99 | |
XM_006247272.1 | XP_006247334.1 | 8 - 9 | 1562 | 99 | ||
GenBank mRNA | BC085881.1 | - | 8 - 9 | 1347 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
