Assay Details
Gene Symbol: Nubp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011891.1 | NP_001011891.1 | 2 - 3 | 161 | 125 | |
GenBank mRNA | BC079386.1 | - | 2 - 3 | 161 | 125 | |
FQ213066.1 | - | 2 - 3 | 175 | 125 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
