Assay Details
Gene Symbol: Akap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053665.1 | NP_446117.1 | 3 - 4 | 1827 | 106 | |
XM_006247067.2 | XP_006247129.1 | 4 - 5 | 2000 | 106 | ||
XM_006247068.1 | XP_006247130.1 | 4 - 5 | 1996 | 106 | ||
XM_008767930.1 | XP_008766152.1 | 4 - 5 | 2055 | 106 | ||
GenBank mRNA | AF068202.1 | - | 3 - 4 | 1827 | 106 | |
BC081753.1 | - | 5 - 6 | 2079 | 106 | ||
BC097298.1 | - | 6 - 7 | 2171 | 106 | ||
BC129090.1 | - | 6 - 7 | 2150 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
