Assay Details
Gene Symbol: Gabrg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006246125.2 | XP_006246187.1 | 8 - 9 | 1691 | 101 | |
GenBank mRNA | AY574252.1 | - | 8 - 9 | 1128 | 101 | |
BC092617.1 | - | 3 - 4 | 391 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
