Assay Details
Gene Symbol: Gabrg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006246125.2 | XP_006246187.1 | 9 - 10 | 1719 | 64 | |
GenBank mRNA | AY574252.1 | - | 9 - 10 | 1156 | 64 | |
BC092617.1 | - | 4 - 5 | 419 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
