Assay Details
Gene Symbol: Unkl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006246059.2 | XP_006246121.1 | 2 - 3 | 2414 | 103 | |
XM_008767609.1 | XP_008765831.1 | 11 - 12 | 1384 | 103 | ||
GenBank mRNA | FQ211597.1 | - | 11 - 12 | 1351 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
