Assay Details
Gene Symbol: Epx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107037.1 | NP_001100507.1 | 6 - 7 | 914 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
