Assay Details
Gene Symbol: Acbd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012013.1 | NP_001012013.1 | 8 - 9 | 118 | ||
XM_006247508.2 | XP_006247570.1 | 9 - 10 | 1144 | 115 | ||
XM_006247510.2 | XP_006247572.1 | 9 - 10 | 118 | |||
XM_006247511.2 | XP_006247573.1 | 9 - 10 | 1135 | 115 | ||
XM_008768287.1 | XP_008766509.1 | 9 - 10 | 118 | |||
GenBank mRNA | BC076387.1 | - | 8 - 9 | 118 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
