Assay Details
Gene Symbol: Pitpna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017231.1 | NP_058927.1 | 3 - 4 | 394 | 88 | |
GenBank mRNA | BC070945.1 | - | 3 - 4 | 422 | 88 | |
M25758.1 | - | 3 - 4 | 394 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
