Assay Details
Gene Symbol: Dhrs7c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271597.1 | NP_001258526.1 | 3 - 4 | 418 | 66 | |
NM_001271598.1 | NP_001258527.1 | 3 - 4 | 418 | 66 | ||
GenBank mRNA | FM120380.1 | - | 3 - 4 | 418 | 66 | |
FN805189.1 | - | 3 - 4 | 373 | 66 | ||
FQ214628.1 | - | 3 - 4 | 399 | 66 | ||
FQ214964.1 | - | 3 - 4 | 398 | 66 | ||
FQ215164.1 | - | 3 - 4 | 399 | 66 | ||
FQ215595.1 | - | 3 - 4 | 399 | 66 | ||
FQ216481.1 | - | 3 - 4 | 399 | 66 | ||
FQ216493.1 | - | 3 - 4 | 399 | 66 | ||
FQ216936.1 | - | 3 - 4 | 399 | 66 | ||
FQ224852.1 | - | 3 - 4 | 399 | 66 | ||
FQ224951.1 | - | 3 - 4 | 398 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
