Assay Details
Gene Symbol: Nme4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109478.1 | NP_001102948.1 | 4 - 5 | 451 | 75 | |
GenBank mRNA | CB724692.1 | - | 3 - 4 | 334 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
