Assay Details
Gene Symbol: Decr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171996.3 | NP_741993.1 | 5 - 6 | 553 | 70 | |
XM_006246024.2 | XP_006246086.1 | 6 - 7 | 753 | 70 | ||
GenBank mRNA | AF044574.1 | - | 5 - 6 | 475 | 70 | |
BC070959.1 | - | 5 - 6 | 554 | 70 | ||
FQ218511.1 | - | 5 - 6 | 528 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
