Assay Details
Gene Symbol: Milr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021585.1 | NP_067596.1 | 3 - 4 | 503 | 95 | |
GenBank mRNA | U39546.1 | - | 3 - 4 | 503 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
