Assay Details
Gene Symbol: Slc13a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170668.1 | NP_733768.1 | 3 - 4 | 389 | 104 | |
XM_006246602.2 | XP_006246664.1 | 4 - 5 | 1615 | 104 | ||
GenBank mRNA | AF522186.1 | - | 3 - 4 | 389 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
