Assay Details
Gene Symbol: Ipo5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006252478.2 | XP_006252540.1 | 7 - 8 | 760 | 117 | |
XM_224534.8 | XP_224534.7 | 8 - 9 | 1006 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
