Assay Details
Gene Symbol: Smarce1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008768051.1 | XP_008766273.1 | 3 - 4 | 492 | 58 | |
GenBank mRNA | BC091314.1 | - | 3 - 4 | 206 | 58 | |
EU327029.1 | - | 3 - 4 | 208 | 58 |
Gene Symbol: LOC103694314 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008772543.1 | XP_008770765.1 | 1 | 2025 | 58 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC091314.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
