Assay Details
Gene Symbol: Smarce1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024993.1 | NP_001020164.1 | 7 - 8 | 714 | 108 | |
XM_008768051.1 | XP_008766273.1 | 7 - 8 | 1056 | 108 | ||
GenBank mRNA | BC091314.1 | - | 7 - 8 | 770 | 108 | |
BC092210.1 | - | 7 - 8 | 714 | 108 | ||
FQ213000.1 | - | 7 - 8 | 702 | 108 |
Gene Symbol: LOC103694314 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008772543.1 | XP_008770765.1 | 1 | 108 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
