Assay Details
Gene Symbol: Lypd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109632.1 | NP_001103102.1 | 4 - 5 | 347 | 75 | |
XM_006246402.2 | XP_006246464.1 | 3 - 4 | 465 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
