Assay Details
Gene Symbol: Sap30bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108305.1 | NP_001101775.1 | 10 - 11 | 783 | 118 | |
GenBank mRNA | BC169106.1 | - | 752 | 118 | ||
FQ220147.1 | - | 10 - 11 | 780 | 118 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P121995 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 53778 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 595428 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 717185 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 265252 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709059 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819829 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 581414 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 371493 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 928643 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 878571 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 195373 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939151 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 87220 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 773084 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434828 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 583618 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872376 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231923 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 491043 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 578727 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63211 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 500093 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565587 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106877 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381644 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150534 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 92861 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 111374 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P121995 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P121995 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P121995 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P110445 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 65245 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 26121 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10739717 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10739714 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10739718 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10739710 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10739712 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10739711 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10739716 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10739720 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10739713 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10739715 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10739719 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10739709 | - | - |
Rat Genome 230 2.0 Array | 1373973_at | - | - |
Rat Genome 230 2.0 Array | 1373973_PM_at | - | - |
Rat Genome 230 2.0 Array | 1383441_PM_at | - | - |
Rat Genome 230 2.0 Array | 1383441_at | - | - |
Affymetrix HT Rat Focus Array | 1373973_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1383441_at | - | - |
Affymetrix HT Rat Focus Array | 1383441_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1373973_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1000017616.rn.1 | TC1000001953.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1000017617.rn.1 | TC1000001953.rn.1 | RTA TC |
Rat Expression Set 230 | 1373973_at | - | - |
Rat Expression Set 230 | 1373973_PM_at | - | - |
Rat Expression Set 230 | 1383441_at | - | - |
Rat Expression Set 230 | 1383441_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10739709 | - | - |
Rat Genome U34 Set | rc_AI169082_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21133239 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1358021 | - | - |
