Assay Details
Gene Symbol: Aatf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053720.1 | NP_446172.1 | 11 - 12 | 1569 | 90 | |
GenBank mRNA | AJ238717.3 | - | 11 - 12 | 1569 | 90 | |
BC078769.1 | - | 11 - 12 | 1546 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
