Assay Details
Gene Symbol: Alox15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031010.2 | NP_112272.2 | 5 - 6 | 653 | 134 | |
XM_006246823.2 | XP_006246885.1 | 7 - 8 | 934 | 134 | ||
XM_006246824.2 | XP_006246886.1 | 6 - 7 | 829 | 134 | ||
GenBank mRNA | L06040.1 | - | 5 - 6 | 666 | 134 | |
S69383.1 | - | 5 - 6 | 653 | 134 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
