Assay Details
Gene Symbol: Map2k3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100674.1 | NP_001094144.1 | 9 - 10 | 931 | 89 | |
GenBank mRNA | BC160839.1 | - | 9 - 10 | 801 | 89 | |
FQ231052.1 | - | 3 - 4 | 261 | 89 |
Gene Symbol: LOC103689956 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008767884.1 | XP_008766106.1 | 9 - 10 | 985 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
