Assay Details
Gene Symbol: Map2k6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053703.3 | NP_446155.1 | 10 - 11 | 991 | 116 | |
GenBank mRNA | AF369384.1 | - | 10 - 11 | 991 | 116 | |
BC087004.1 | - | 10 - 11 | 1091 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
