Assay Details
Gene Symbol: Cdc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108298.1 | NP_001101768.1 | 11 - 12 | 1783 | 140 | |
XM_006247354.2 | XP_006247416.1 | 11 - 12 | 1785 | 140 | ||
XM_006247355.2 | XP_006247417.1 | 11 - 12 | 1783 | 140 | ||
GenBank mRNA | FQ233913.1 | - | 2 - 3 | 284 | 140 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
