Assay Details
Gene Symbol: Cdc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108298.1 | NP_001101768.1 | 2 - 3 | 363 | 132 | |
XM_006247354.2 | XP_006247416.1 | 2 - 3 | 365 | 132 | ||
XM_006247355.2 | XP_006247417.1 | 2 - 3 | 366 | 132 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
