Assay Details
Gene Symbol: Tp53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030989.3 | NP_112251.2 | 6 - 7 | 908 | 70 | |
XM_006246594.2 | XP_006246656.1 | 7 - 8 | 1337 | 70 | ||
XM_006246595.2 | XP_006246657.1 | 6 - 7 | 1694 | 70 | ||
XM_008767773.1 | XP_008765995.1 | 6 - 7 | 915 | 70 | ||
GenBank mRNA | BC081788.1 | - | 6 - 7 | 837 | 70 | |
BC098663.1 | - | 6 - 7 | 866 | 70 | ||
X13058.1 | - | 5 - 6 | 800 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
