Assay Details
Gene Symbol: Apoh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009626.1 | NP_001009626.1 | 1 - 2 | 68 | 71 | |
GenBank mRNA | BC088180.1 | - | 1 - 2 | 68 | 71 | |
FQ209491.1 | - | 1 - 2 | 94 | 71 | ||
FQ209740.1 | - | 1 - 2 | 103 | 71 | ||
FQ210182.1 | - | 1 - 2 | 94 | 71 | ||
FQ210311.1 | - | 1 - 2 | 94 | 71 | ||
FQ210756.1 | - | 1 - 2 | 110 | 71 | ||
FQ210813.1 | - | 1 - 2 | 94 | 71 | ||
FQ211328.1 | - | 1 - 2 | 67 | 71 | ||
FQ218445.1 | - | 1 - 2 | 108 | 71 | ||
FQ218454.1 | - | 1 - 2 | 94 | 71 | ||
FQ218597.1 | - | 1 - 2 | 90 | 71 | ||
FQ218599.1 | - | 1 - 2 | 94 | 71 | ||
FQ218637.1 | - | 1 - 2 | 94 | 71 | ||
FQ218785.1 | - | 1 - 2 | 110 | 71 | ||
FQ218859.1 | - | 1 - 2 | 67 | 71 | ||
FQ218865.1 | - | 1 - 2 | 94 | 71 | ||
FQ218961.1 | - | 1 - 2 | 94 | 71 | ||
FQ219138.1 | - | 1 - 2 | 159 | 71 | ||
FQ219328.1 | - | 1 - 2 | 94 | 71 | ||
FQ219380.1 | - | 1 - 2 | 95 | 71 | ||
FQ219481.1 | - | 1 - 2 | 107 | 71 | ||
FQ219537.1 | - | 1 - 2 | 91 | 71 | ||
FQ219580.1 | - | 1 - 2 | 108 | 71 | ||
FQ219601.1 | - | 1 - 2 | 107 | 71 | ||
FQ219674.1 | - | 1 - 2 | 94 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
