Assay Details
Gene Symbol: Apoh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009626.1 | NP_001009626.1 | 5 - 6 | 607 | 73 | |
GenBank mRNA | AM071386.1 | - | 5 - 6 | 566 | 73 | |
BC088180.1 | - | 5 - 6 | 607 | 73 | ||
FQ209412.1 | - | 5 - 6 | 646 | 73 | ||
FQ209491.1 | - | 5 - 6 | 633 | 73 | ||
FQ209740.1 | - | 5 - 6 | 642 | 73 | ||
FQ210182.1 | - | 5 - 6 | 633 | 74 | ||
FQ210311.1 | - | 5 - 6 | 633 | 73 | ||
FQ210756.1 | - | 5 - 6 | 649 | 73 | ||
FQ210813.1 | - | 5 - 6 | 633 | 73 | ||
FQ218445.1 | - | 5 - 6 | 647 | 73 | ||
FQ218597.1 | - | 5 - 6 | 629 | 73 | ||
FQ218599.1 | - | 5 - 6 | 633 | 73 | ||
FQ218785.1 | - | 5 - 6 | 649 | 73 | ||
FQ218865.1 | - | 5 - 6 | 633 | 73 | ||
FQ218961.1 | - | 5 - 6 | 633 | 73 | ||
FQ219138.1 | - | 5 - 6 | 698 | 73 | ||
FQ219328.1 | - | 5 - 6 | 633 | 73 | ||
FQ219380.1 | - | 5 - 6 | 634 | 73 | ||
FQ219481.1 | - | 5 - 6 | 646 | 73 | ||
FQ219537.1 | - | 5 - 6 | 630 | 73 | ||
FQ219580.1 | - | 5 - 6 | 647 | 73 | ||
FQ219674.1 | - | 5 - 6 | 633 | 73 | ||
X15551.1 | - | 5 - 6 | 462 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
