Assay Details
Gene Symbol: Mrpl38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009369.2 | NP_001009369.2 | 1 - 2 | 82 | 104 | |
GenBank mRNA | BC087096.1 | - | 1 - 2 | 79 | 104 | |
CO567099.1 | - | 1 - 2 | 82 | 104 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC087096.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
