Assay Details
Gene Symbol: Cldn10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106058.1 | NP_001099528.1 | 2 - 3 | 633 | 58 | |
XM_006252451.2 | XP_006252513.1 | 1 - 2 | 366 | 58 | ||
GenBank mRNA | BC158829.1 | - | 1 - 2 | 236 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
