Assay Details
Gene Symbol: Cldn10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106058.1 | NP_001099528.1 | 3 - 4 | 797 | 143 | |
XM_006252450.2 | XP_006252512.1 | 2 - 3 | 638 | 143 | ||
XM_006252451.2 | XP_006252513.1 | 2 - 3 | 530 | 143 | ||
GenBank mRNA | BC158829.1 | - | 2 - 3 | 400 | 143 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
