Assay Details
Gene Symbol: Akap11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012773.2 | NP_036905.2 | 5 - 6 | 636 | 65 | |
XM_006252344.1 | XP_006252406.1 | 6 - 7 | 810 | 65 | ||
XM_008770886.1 | XP_008769108.1 | 6 - 7 | 1702 | 65 | ||
XM_008770887.1 | XP_008769109.1 | 6 - 7 | 892 | 65 | ||
XM_008770888.1 | XP_008769110.1 | 6 - 7 | 892 | 65 | ||
XM_008770889.1 | XP_008769111.1 | 6 - 7 | 892 | 65 | ||
XM_008770890.1 | XP_008769112.1 | 6 - 7 | 892 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
