Assay Details
Gene Symbol: Bora | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013997.1 | NP_001014019.1 | 1 - 2 | 250 | 59 | |
XM_008770868.1 | XP_008769090.1 | 1 - 2 | 434 | 59 | ||
GenBank mRNA | BC088204.1 | - | 1 - 2 | 250 | 59 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC088204.1, NM_001013997.1, XM_008770868.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
