Assay Details
Gene Symbol: Lclat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008764536.1 | XP_008762758.1 | 4 - 5 | 8115 | 138 | |
XM_008764537.1 | XP_008762759.1 | 4 - 5 | 665 | 138 | ||
XM_008764538.1 | XP_008762760.1 | 4 - 5 | 655 | 138 | ||
XM_008764539.1 | XP_008762761.1 | 4 - 5 | 8115 | 138 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
