Assay Details
Gene Symbol: Gemin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053389.1 | NP_445841.1 | 4 - 5 | 452 | 115 | |
GenBank mRNA | AF176072.1 | - | 4 - 5 | 452 | 115 | |
BC098662.1 | - | 4 - 5 | 395 | 115 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
