Assay Details
Gene Symbol: Arhgap5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008764656.1 | XP_008762878.1 | 1 - 2 | 3739 | 136 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
