Assay Details
Gene Symbol: Rnaseh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013097.2 | NP_001013115.1 | 4 - 5 | 565 | 67 | |
NM_001286938.1 | NP_001273867.1 | 4 - 5 | 565 | 67 | ||
GenBank mRNA | BC091209.1 | - | 4 - 5 | 540 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
